Läs mer
[3-(3,4-Dimethoxyphenyl)propyl](methyl)amine Hydrochloride, TRC
Produktkod. 30336505
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30336505 25 mg
1 options
Produktkod. 30336505 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB43217025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material [3-(3,4-Dimethoxyphenyl)propyl](methyl)amine Hydrochloride
CAS 83979-31-1
Molekylformel C12H19NO2 . HCl
Kvantitet 25 mg
Synonym Benzenepropanamine, 3,4-dimethoxy-N-methyl-, hydrochloride (1:1) (ACI), Benzenepropanamine, 3,4-dimethoxy-N-methyl-, hydrochloride (9CI), [3-(3,4-Dimethoxyphenyl)propyl](methyl)amine hydrochloride, 3,4-Dimethoxy-N-methylbenzenepropanamine hydrochloride
InChI formel InChI=1S/C12H19NO2.ClH/c1-13-8-4-5-10-6-7-11(14-2)12(9-10)15-3;/h6-7,9,13H,4-5,8H2,1-3H3;1H
LEDER Cl.CNCCCc1ccc(OC)c(OC)c1
IUPAC-namn 3-(3,4-dimethoxyphenyl)-N-methylpropan-1-amine;hydrochloride
Molekylvikt (g/mol) 245.74
Formel vikt 245.1183