Läs mer
3,3',5,5'-Tetraiodo Thyroformic Acid (>90%), TRC
Produktkod. 30010868
Change view
Kvantitet:
2.5 mg
25 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30010868 2.5 mg 2.5mg
30012733 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30010868 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT2961502.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2055-97-2
Molekylformel C13 H6 I4 O4
Molekylvikt (g/mol) 733.8
Synonym 4-(4-Hydroxy-3,5-diiodophenoxy)-3,5-diiodobenzoic Acid, Benzoic acid, 4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodo-, 3,3',5,5'-Tetraiodothyroformic acid, 3,5-Diiodo-4-(3',5'-diiodo-4'-hydroxyphenoxy)benzoic acid, 4-(4-Hydroxy-3,5-diiodophenoxy)-3,5-diiodobenzoic acid, TFA 4, Tetraform, Tetraform (thyroid hormone analog), Tetraiodothyroformic acid, Levothyroxine EP impurity H
IUPAC-namn 4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodobenzoic acid
LEDER OC(=O)c1cc(I)c(Oc2cc(I)c(O)c(I)c2)c(I)c1
Specifikationer
Kemiskt namn eller material 3,3',5,5'-Tetraiodo Thyroformic Acid
Anteckningar om renhetsgrad HPLC
Kvantitet 2.5 mg
InChI formel InChI=1S/C13H6I4O4/c14-7-3-6(4-8(15)11(7)18)21-12-9(16)1-5(13(19)20)2-10(12)17/h1-4,18H,(H,19,20)
Formel vikt 733.6445
Procent renhet >95%