Läs mer
3,3',5,5'-Tetraiodothyronamine Hydrochloride, TRC
Produktkod. 30423168
Change view
Kvantitet:
2.5 mg
25 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30423168 2.5 mg 2.5mg
30415079 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30423168 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT2961802.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 788824-71-5
Molekylformel C14H11I4NO2.HCl
Molekylvikt (g/mol) 769.32
Synonym 4-[4-(2-Aminoethyl)-2,6-diiodophenoxy]-2,6-diiodophenol Hydrochloride
IUPAC-namn 4-[4-(2-aminoethyl)-2,6-diiodophenoxy]-2,6-diiodophenol;hydrochloride
LEDER Cl.NCCc1cc(I)c(Oc2cc(I)c(O)c(I)c2)c(I)c1
Specifikationer
Kemiskt namn eller material 3,3',5,5'-Tetraiodothyronamine Hydrochloride
Kvantitet 2.5 mg
InChI formel InChI=1S/C14H11I4NO2.ClH/c15-9-5-8(6-10(16)13(9)20)21-14-11(17)3-7(1-2-19)4-12(14)18;/h3-6,20H,1-2,19H2;1H
Formel vikt 768.6735