Läs mer
3-[3-Acetyl-4-[(2RS)-3-(diethylamino)-2-hydroxypropoxy]phenyl]-1,1-diethylurea, Mikromol™
Produktkod. 30457903
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30457903 4 x 25 mg
17959754 25 mg
17939174 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30457903 Leverantör Mikromol Leverantörsnummer MM0128.1000254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3-[3-Acetyl-4-[(2RS)-3-(diethylamino)-2-hydroxypropoxy]phenyl]-1,1-diethylurea, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-[3-Acetyl-4-[(2RS)-3-(diethylamino)-2-hydroxypropoxy]phenyl]-1,1-diethylurea
CAS 2727065-34-9
Analyt- eller komponentnamn 3-[3-Acetyl-4-[(2RS)-3-(diethylamino)-2-hydroxypropoxy]phenyl]-1,1-diethylurea
Frakt skick Room Temperature
Molekylformel C20 H33 N3 O4
Certifikat och efterlevnad ISO 17025
Synonym Celiprolol Hydrochloride Imp. D (EP), Celiprolol Imp. D (EP), 3-[3-Acetyl-4-[(2RS)-3-(diethylamino)-2-hydroxypropoxy]phenyl]-1,1-diethylurea, Celiprolol Hydrochloride Impurity D, Celiprolol Impurity D
InChI formel InChI=1S/C25H31NO3.C2H2O4/c1-4-7-11-23-24(21-10-8-9-12-22(21)29-23)25(27)19-13-15-20(16-14-19)28-18-17-26(5-2)6-3;3-1(4)2(5)6/h8-10,12-16H,4-7,11,17-18H2,1-3H3;(H,3,4)(H,5,6)
Lösningstyp Neat
LEDER CCN(CC)CC(O)COc1ccc(NC(=O)N(CC)CC)cc1C(=O)C
IUPAC-namn 3-[3-acetyl-4-[3-(diethylamino)-2-hydroxypropoxy]phenyl]-1,1-diethylurea
Molekylvikt (g/mol) 379.49
Kvantitet 4 x 25 mg
Formel vikt 379.2471 g/mol