Läs mer
3-[3-Acetyl-4-[[(RS)-oxiranyl]-methoxy]phenyl]-1,1-diethylurea, Mikromol™
Produktkod. 30050586
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30050586 25 mg
30049600 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30050586 Leverantör Mikromol Leverantörsnummer MM0128.060025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3-[3-Acetyl-4-[[(RS)-oxiranyl]-methoxy]phenyl]-1,1-diethylurea
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-[3-Acetyl-4-[((RS)-oxiranyl)-methoxy]phenyl]-1,1-diethylurea
CAS 56980-98-4
Analyt- eller komponentnamn 3-[3-Acetyl-4-[((RS)-oxiranyl)-methoxy]phenyl]-1,1-diethylurea
Frakt skick Ice pack (-20°C)
Molekylformel C16 H22 N2 O4
Certifikat och efterlevnad ISO 9001
Synonym Celiprolol Hydrochloride Imp. G (EP), Celiprolol Imp. G (EP), 3-[3-Acetyl-4-[[(RS)-oxiranyl]-methoxy]phenyl]-1,1-diethylurea, Celiprolol Hydrochloride Impurity G, Celiprolol Impurity G
InChI formel InChI=1S/C16H22N2O4/c1-4-18(5-2)16(20)17-12-6-7-15(14(8-12)11(3)19)22-10-13-9-21-13/h6-8,13H,4-5,9-10H2,1-3H3,(H,17,20)
LEDER CCN(CC)C(=O)Nc1ccc(OCC2CO2)c(c1)C(=O)C
IUPAC-namn 3-[3-acetyl-4-(oxiran-2-ylmethoxy)phenyl]-1,1-diethylurea
Molekylvikt (g/mol) 306.36
Kvantitet 25 mg
Formel vikt 306.158 g/mol