Learn More
Alfa Aesar™ 3-(3-Chlorophenyl)-1-phenyl-2-thiohydantoin, 95%
Brand:  Alfa Aesar™ H64397.06
Additional Details : Weight : 0.00500kg
Quantity 5g
 View more versions of this product
Product Code. 15471589
  • 6382.67 SEK / 5g
Estimated Shipment date
from Supplier 06-03-2018
Add to basket
Specifications
Chemical Identifiers
3-(3-Chlorophenyl)-1-phenyl-2-thiohydantoin
302.78
MFCD27665276
3-(3-Chlorophenyl)-1-phenyl-2-thioxo-4-imidazolidinone
Specifications
3-(3-Chlorophenyl)-1-phenyl-2-thiohydantoin
Solid
302.78
3-(3-Chlorophenyl)-1-phenyl-2-thioxo-4-imidazolidinone
5g
95%
MFCD27665276