Learn More
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate, 98%, BLD Pharm™
Product Code. 31450464
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
10 g
25 g
100 g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31450464 250 mg
31438779 1 g
31442698 25 g
31438954 5 g
31443470 100 g
31434101 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 31450464 Supplier BLD Pharm Supplier No. BD120556250mg
 Restricted Item
Beräknad leverans: 25-11-2026
Specifications
CAS 75621-03-3
Molecular Formula C32H58N2O7S
MDL Number MFCD00081083
Quantity 250 mg
SMILES C[C@H](CCC(NCCC[N+](C)(C)CCCS(=O)([O-])=O)=O)[C@H]1CC[C@@]2([H])[C@]3([H])[C@H](O)C[C@]4([H])C[C@H](O)CC[C@]4(C)[C@@]3([H])C[C@H](O)[C@]12C
Molecular Weight (g/mol) 614.88
Percent Purity 98%
Chemical Name or Material 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate