missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 3,3'-Diaminobenzidine, 99%

Catalog Number p-100402
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
91-95-2
C12H14N4
214.27
MFCD00007725
HSTOKWSFWGCZMH-UHFFFAOYSA-N
3,3'-diaminobenzidine, 3,3',4,4'-tetraaminobiphenyl, 3,3'-diaminobenzidene, 3,3',4,4'-diphenyltetramine, 3,3',4,4'-tetraminobiphenyl, 3,3',4,4'-tetraaminodiphenyl, 1,1'-biphenyl-3,3',4,4'-tetramine, biphenyl-3,3',4,4'-tetrayltetraamine, unii-2rv4t6khqi, 4-3,4-diaminophenyl benzene-1,2-diamine
7071
4-(3,4-diaminophenyl)benzene-1,2-diamine
C1=CC(=C(C=C1C2=CC(=C(C=C2)N)N)N)N
This item is not returnable. View return policy
91-95-2
C12H14N4
214.27
MFCD00007725
HSTOKWSFWGCZMH-UHFFFAOYSA-N
3,3'-diaminobenzidine, 3,3',4,4'-tetraaminobiphenyl, 3,3'-diaminobenzidene, 3,3',4,4'-diphenyltetramine, 3,3',4,4'-tetraminobiphenyl, 3,3',4,4'-tetraaminodiphenyl, 1,1'-biphenyl-3,3',4,4'-tetramine, biphenyl-3,3',4,4'-tetrayltetraamine, unii-2rv4t6khqi, 4-3,4-diaminophenyl benzene-1,2-diamine
7071
4-(3,4-diaminophenyl)benzene-1,2-diamine
C1=CC(=C(C=C1C2=CC(=C(C=C2)N)N)N)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 176.0°C to 180.0°C
Flash Point >200°C
Assay Percent Range 99%
Packaging Glass bottle
Quantity 5 g
Beilstein 13, 340
Solubility Information Solubility in water: 0.55g/L in water (20°C); 25g/L in water (95°C).
Formula Weight 214.27
Percent Purity 99%
Chemical Name or Material 3, 3'-Diaminobenzidine, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 3,3'-Diaminobenzidine
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H350-May cause cancer.
  • H341-Suspected of causing genetic defects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'