Läs mer
3,3’-Diaminobenzidine Tetrahydrochloride, TRC
Produktkod. 30349964
Change view
Kvantitet:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30349964 5 g
30354806 10 g
30364188 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30349964 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4161505G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 7411-49-6
Molekylformel C12 H14 N4 . 4 Cl H
Molekylvikt (g/mol) 360.11
IUPAC-namn 4-(3,4-diaminophenyl)benzene-1,2-diamine;tetrahydrochloride
LEDER Cl.Cl.Cl.Cl.Nc1ccc(cc1N)c2ccc(N)c(N)c2
Specifikationer
Kemiskt namn eller material 3,3'-Diaminobenzidine Tetrahydrochloride
Kvantitet 5 g
InChI formel InChI=1S/C12H14N4.4ClH/c13-9-3-1-7(5-11(9)15)8-2-4-10(14)12(16)6-8;;;;/h1-6H,13-16H2;4*1H
Formel vikt 358.0286