Läs mer
3,3'-Diaminobenzidine, TRC
Produktkod. 30367088
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30367088 500 mg
1 options
Produktkod. 30367088 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD415020500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3,3'-Diaminobenzidine, tetrahydrochloride, dihydrate
CAS 167684-17-5
Molekylformel C12 H14 N4 . 4[H Cl] . 2[H2 O]
Kvantitet 500 mg
Synonym 3,3'-Diaminobenzidine, [1,1′-Biphenyl]-3,3′,4,4′-tetramine, tetrahydrochloride, dihydrate (9CI)
InChI formel InChI=1S/C12H14N4.4ClH.2H2O/c13-9-3-1-7(5-11(9)15)8-2-4-10(14)12(16)6-8;;;;;;/h1-6H,13-16H2;4*1H;2*1H2
LEDER O.O.Cl.Cl.Cl.Cl.Nc1ccc(cc1N)c2ccc(N)c(N)c2
IUPAC-namn 4-(3,4-diaminophenyl)benzene-1,2-diamine;dihydrate;tetrahydrochloride
Molekylvikt (g/mol) 396.14
Formel vikt 394.0497