Läs mer
3,3'-Dichlorobisphenol A, TRC
Produktkod. 17903405
Change view
Kvantitet:
1 mg
5 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17903405 25 mg
17900685 1 mg
17902905 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17903405 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD43355025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 79-98-1
Molekylformel C15 H14 Cl2 O2
Molekylvikt (g/mol) 297.18
Synonym 4,4'-(1-Methylethylidene)bis[2-chlorophenol], 2,2-Bis(3-chloro-4-hydroxyphenyl) propane, 2,2-Di(4-hydroxy-3-chlorophenyl)propane, 2,2'-Dichlorobisphenol A, 3,3'-Dichlorodian, NSC 18247
IUPAC-namn 2-chloro-4-[1-(3-chloro-4-hydroxy-phenyl)-1-methyl-ethyl]phenol
LEDER CC(C)(c1ccc(O)c(Cl)c1)c2ccc(O)c(Cl)c2
Specifikationer
Kemiskt namn eller material 3,3'-Dichlorobisphenol A
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C15H14Cl2O2/c1-15(2,9-3-5-13(18)11(16)7-9)10-4-6-14(19)12(17)8-10/h3-8,18-19H,1-2H3
Formel vikt 296.0371
Procent renhet >95%