Läs mer
3,3-Diethoxypropionitrile, TRC
Produktkod. 30376195
Change view
Kvantitet:
50 g
Produktkod. Kvantitet
30376195 50 g
1 options
Produktkod. 30376195 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD44210050G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3,3-Diethoxypropionitrile
CAS 2032-34-0
Molekylformel C7H13NO2
Kvantitet 50 g
Synonym 3,3-Diethoxypropanenitrile, Cyanoacetaldehyde Diethyl Acetal, Malonaldehydenitrile Diethyl Acetal
InChI formel InChI=1S/C7H13NO2/c1-3-9-7(5-6-8)10-4-2/h7H,3-5H2,1-2H3
LEDER CCOC(CC#N)OCC
Molekylvikt (g/mol) 143.18
Formel vikt 143.0946