Läs mer
3,3'-Dihydroxyazoxybenzene, Mikromol™
Produktkod. 30460842
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30460842 4 x 25 mg
30051964 25 mg
30049090 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30460842 Leverantör Mikromol Leverantörsnummer MM1645.0100254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3,3'-Dihydroxyazoxybenzene, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3,3'-Dihydroxyazoxybenzene
CAS 17540-51-1
Analyt- eller komponentnamn 3,3'-Dihydroxyazoxybenzene
Frakt skick Room Temperature
Molekylformel C12 H10 N2 O3
Certifikat och efterlevnad ISO 9001
Synonym Phenol, 3,3'-azoxybis- (9CI)
InChI formel InChI=1S/C12H10N2O3/c15-11-5-1-3-9(7-11)13-14(17)10-4-2-6-12(16)8-10/h1-8,15-16H/b14-13-
Lösningstyp Neat
LEDER Oc1cccc(c1)N=[N+]([O-])c2cccc(O)c2
IUPAC-namn (Z)-(3-hydroxyphenyl)-(3-hydroxyphenyl)imino-oxidoazanium
Molekylvikt (g/mol) 230.22
Kvantitet 4 x 25 mg
Formel vikt 230.0691 g/mol