missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3'-Diindolylmethane, 96%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
Specifications
Melting Point | 164.0°C to 168.0°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 95% min. (HPLC) |
Quantity | 5 g |
Synonym | 3,3'-diindolylmethane, di 1h-indol-3-yl methane, diindolylmethane, 3,3'-methylenediindole, 3,3'-methylenebis-1h-indole, 1h-indole, 3,3'-methylenebis, dim, 3-diindolyl methane, arundine, 3,3'-methylenebis 1h-indole |
InChI Key | VFTRKSBEFQDZKX-UHFFFAOYSA-N |
SMILES | C1=CC=C2C(=C1)C(=CN2)CC3=CNC4=CC=CC=C43 |
IUPAC Name | 3-(1H-indol-3-ylmethyl)-1H-indole |
PubChem CID | 3071 |
Formula Weight | 246.31 |
Show More |
Safety and Handling
Product Identifier
- 3,3'-Diindolylmethane
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H413-May cause long lasting harmful effects to aquatic life.
- H315-Causes skin irritation.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'