missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethoxypropanenitrile, 90%, Thermo Scientific Chemicals

Catalog Number p-4812874
Click to view available options
Quantity:
25 mL
100 mL
This item is not returnable. View return policy
57597-62-3
C5H9NO2
115.13
MFCD00044797
JYHSJQNYYLGMEI-UHFFFAOYSA-N
93731
3,3-dimethoxypropanenitrile
COC(CC#N)OC
This item is not returnable. View return policy
57597-62-3
C5H9NO2
115.13
MFCD00044797
JYHSJQNYYLGMEI-UHFFFAOYSA-N
93731
3,3-dimethoxypropanenitrile
COC(CC#N)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.0260g/mL
Boiling Point 195°C
Flash Point 83°C
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula (CH3O)2CHCH2CN
Refractive Index 1.4130 to 1.415
Quantity 25 mL
Specific Gravity 1.026
Synonym 3,3-dimethoxypropionitrile, propanenitrile, 3,3-dimethoxy, 3,3-dimethoxypropiononitrile, 3,3-dimethoxy propionitrile, 3,3-dimethoxy-propionitrile, 3,3-dimethoxy-propanenitrile, 3,3-dimethoxypropanitrile, acmc-209lz7, #, propanenitrile,3,3-dimethoxy
Solubility Information Solubility in water: soluble
Formula Weight 115.13
Percent Purity 90%
Chemical Name or Material 3, 3-Dimethoxypropanenitrile
Show More Show Less
compliance-icons
Product Identifier
  • 3,3-Dimethoxy-propanenitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'