missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethyl-1,2-epoxybutane, 95%, Thermo Scientific Chemicals

Catalog Number p-7044528
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
2245-30-9
C6H12O
100.161
MFCD00051590
HEAYDCIZOFDHRM-UHFFFAOYSA-N
3,3-dimethyl-1,2-epoxybutane, tert-butyloxirane, tert-butylethylene oxide, 3,3-dimethylbutene oxide, oxirane, 1,1-dimethylethyl, 3,3-dimethyl-1-butene oxide, 1,2-epoxy-3,3-dimethylbutane, tert.-butyloxirane, tert-butyl oxirane, butane, 1,2-epoxy-3,3-dimethyl
92174
2-tert-butyloxirane
CC(C)(C)C1CO1
This item is not returnable. View return policy
2245-30-9
C6H12O
100.161
MFCD00051590
HEAYDCIZOFDHRM-UHFFFAOYSA-N
3,3-dimethyl-1,2-epoxybutane, tert-butyloxirane, tert-butylethylene oxide, 3,3-dimethylbutene oxide, oxirane, 1,1-dimethylethyl, 3,3-dimethyl-1-butene oxide, 1,2-epoxy-3,3-dimethylbutane, tert.-butyloxirane, tert-butyl oxirane, butane, 1,2-epoxy-3,3-dimethyl
92174
2-tert-butyloxirane
CC(C)(C)C1CO1

3,3-Dimethyl-1,2-epoxybutane is used as pharm and intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3-Dimethyl-1,2-epoxybutane is used as pharm and intermediates.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 0.822
Boiling Point 95°C to 96°C
Flash Point −1°C (30°F)
Odor Recognizable
Refractive Index 1.401
Quantity 5 g
UN Number UN1993
Beilstein 102478
Formula Weight 100.16
Percent Purity 95%
Chemical Name or Material 3,3-Dimethyl-1,2-epoxybutane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'