Läs mer
3,3'-Dimethyl-5,5'-bis(1-methylethyl)-[2,2'-binaphthalene]-1,1',6,6',7,7'-hexol Hexaacetate, TRC
Produktkod. 30366224
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30366224 100 mg
1 options
Produktkod. 30366224 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD105200100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3,3'-Dimethyl-5,5'-bis(1-methylethyl)-[2,2'-binaphthalene]-1,1',6,6',7,7'-hexol Hexaacetate
CAS 105201-55-6
Molekylformel C40H42O12
Kvantitet 100 mg
Synonym NSC 159451, 5,5'-Diisopropyl-3,3'-dimethyl-[2,2'-binaphthalene]-1,1',6,6',7,7'-hexayl Hexaacetate
InChI formel InChI=1S/C40H42O12/c1-17(2)33-27-13-19(5)35(37(49-23(9)43)29(27)15-31(47-21(7)41)39(33)51-25(11)45)36-20(6)14-28-30(38(36)50-24(10)44)16-32(48-22(8)42)40(52-26(12)46)34(28)18(3)4/h13-18H,1-12H3
LEDER CC(C)c1c(OC(=O)C)c(OC(=O)C)cc2c(OC(=O)C)c(c(C)cc12)c3c(C)cc4c(C(C)C)c(OC(=O)C)c(OC(=O)C)cc4c3OC(=O)C
IUPAC-namn [3,8-diacetyloxy-6-methyl-4-propan-2-yl-7-(1,6,7-triacetyloxy-3-methyl-5-propan-2-ylnaphthalen-2-yl)naphthalen-2-yl] acetate
Molekylvikt (g/mol) 714.25