missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(3-Pyridyl)benzeneboronic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
It is used as an OLED material. Phenylboronic acids are efficient phenylating agents in copper promoted CN and CO bond cross-coupling reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as an OLED material. Phenylboronic acids are efficient phenylating agents in copper promoted CN and CO bond cross-coupling reactions.
Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
It is used as an OLED material. Phenylboronic acids are efficient phenylating agents in copper promoted CN and CO bond cross-coupling reactions.
Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Specifications
Color | White |
Quantity | 1 g |
Beilstein | 20520996 |
IUPAC Name | (3-pyridin-3-ylphenyl)boronic acid |
Formula Weight | 199.02 |
Percent Purity | 98% |
Physical Form | Powder |
Chemical Name or Material | 3-(3-Pyridyl)benzeneboronic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'