Learn More
3,3-Tetramethyleneglutaric acid, 98+%, Thermo Scientific™
Product Code. 11470681
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
Product Code. Quantity Unit Size
11470681 100 g 100g
1 options
Product Code. 11470681 Supplier Thermo Scientific Alfa Aesar Supplier No. B20146.22
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Este artículo no se puede devolver. Vea la política de devoluciones
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Identificadores de productos químicos
CAS 16713-66-9
Molecular Formula C9H14O4
Molecular Weight (g/mol) 186.21
MDL Number MFCD00001388
InChI Key FWPVKDFOUXHOKQ-UHFFFAOYSA-N
Synonym 1,1-cyclopentanediacetic acid, 3,3-tetramethyleneglutaric acid, cyclopentane-1,1-diacetic acid, cyclopentanediacetic acid, tetramethyleneglutaric acid, tetramethylene glutaric acid, 2-1-carboxymethyl cyclopentyl acetic acid, 1,1-cyclopentanediaceticacid, chembl80679, 3,3'-tetramethylene glutaric acid
PubChem CID 85558
IUPAC Name 2-[1-(carboxymethyl)cyclopentyl]acetic acid
SMILES OC(=O)CC1(CC(O)=O)CCCC1
Especificaciones
Melting Point 179°C to 181°C
Quantity 100 g
Beilstein 2099523
Formula Weight 186.21
Percent Purity ≥98%
Chemical Name or Material 3,3-Tetramethyleneglutaric acid
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.