Läs mer
3-((3R,4R)-4-Methyl-3-(methyl(7H-pyrrolo[2,3-d]pyrimidin-4-yl)amino)piperidin-1-yl)-3-oxopropanoic Acid, TRC
Produktkod. 30481338
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30481338 50 mg
30479534 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30481338 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM32439550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2328165-36-0
Molekylformel C16H21N5O3
Molekylvikt (g/mol) 331.37
Synonym Tofacitinib impurity K
LEDER C[C@@H]1CCN(C[C@@H]1N(C)c2ncnc3[nH]ccc23)C(=O)CC(=O)O
Specifikationer
Kemiskt namn eller material 3-((3R,4R)-4-Methyl-3-(methyl(7H-pyrrolo[2,3-d]pyrimidin-4-yl)amino)piperidin-1-yl)-3-oxopropanoic Acid
Kvantitet 50 mg
InChI formel InChI=1S/C16H21N5O3/c1-10-4-6-21(13(22)7-14(23)24)8-12(10)20(2)16-11-3-5-17-15(11)18-9-19-16/h3,5,9-10,12H,4,6-8H2,1-2H3,(H,23,24)(H,17,18,19)/t10-,12+/m1/s1