Läs mer
3,4,4',5-Tetrachlorobiphenyl, TRC
Produktkod. 30452825
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30452825 50 mg
30451084 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30452825 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT88998550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 70362-50-4
Molekylformel C12 H6 Cl4
Molekylvikt (g/mol) 291.99
Synonym 1,1'-Biphenyl, 3,4,4',5-tetrachloro-, 3,4,4',5-Tetrachloro-1,1'-biphenyl, 3,4,4',5-TeCB, 3,4,4',5-Tetrachlorobiphenyl, 3,4,5,4'-Tetrachlorobiphenyl, CB 81, PCB 81, Polychlorinated biphenyl-81, TeCB 81, 3,4,4',5-TetraCB
IUPAC-namn 1,2,3-trichloro-5-(4-chlorophenyl)benzene
LEDER Clc1ccc(cc1)c2cc(Cl)c(Cl)c(Cl)c2
Specifikationer
Kemiskt namn eller material PCB No. 81
Kvantitet 50 mg
InChI formel InChI=1S/C12H6Cl4/c13-9-3-1-7(2-4-9)8-5-10(14)12(16)11(15)6-8/h1-6H
Formel vikt 289.9224