Läs mer
3-[4-(4-Chlorobenzoyl)phenoxy]-2-butanone (Fenofibrate Impurity), TRC
Produktkod. 30132399
Change view
Kvantitet:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132399 250 mg
30012296 50 mg
30012398 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30132399 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC364540250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 217636-47-0
Molekylformel C17 H15 Cl O3
Molekylvikt (g/mol) 302.75
Synonym (3RS)-3-[4-(4-Chlorobenzoyl)phenoxy]butan-2-one, Fenofibrate Imp. C (EP)
IUPAC-namn 3-[4-(4-chlorobenzoyl)phenoxy]butan-2-one
LEDER CC(Oc1ccc(cc1)C(=O)c2ccc(Cl)cc2)C(=O)C
Specifikationer
Kemiskt namn eller material (3RS)-3-[4-(4-Chlorobenzoyl)phenoxy]butan-2-one
Anteckningar om renhetsgrad HPLC
Kvantitet 250 mg
InChI formel InChI=1S/C17H15ClO3/c1-11(19)12(2)21-16-9-5-14(6-10-16)17(20)13-3-7-15(18)8-4-13/h3-10,12H,1-2H3
Formel vikt 302.071
Procent renhet >95%