missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trichloropyridazine, 98%, Thermo Scientific Chemicals

Catalog Number 15575866
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
14161-11-6
C4HCl3N2
183.42
GBAOOJAWDCNOGO-UHFFFAOYSA-N
pyridazine, 3,4,5-trichloro, 3,4,5-trichloro-pyridazine, trichloropyridazine, ksc493s5t, gbaoojawdcnogo-uhfffaoysa, pyridazine,3,4,5-trichloro, 3,4,5-trichloro-1,2-diazine
70111
3,4,5-trichloropyridazine
C1=C(C(=C(N=N1)Cl)Cl)Cl
This item is not returnable. View return policy
14161-11-6
C4HCl3N2
183.42
GBAOOJAWDCNOGO-UHFFFAOYSA-N
pyridazine, 3,4,5-trichloro, 3,4,5-trichloro-pyridazine, trichloropyridazine, ksc493s5t, gbaoojawdcnogo-uhfffaoysa, pyridazine,3,4,5-trichloro, 3,4,5-trichloro-1,2-diazine
70111
3,4,5-trichloropyridazine
C1=C(C(=C(N=N1)Cl)Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 54.0°C to 61.0°C
Boiling Point 117.0°C to 118.0°C (14.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 183.42
Percent Purity 98%
Chemical Name or Material 3,4,5-Trichloropyridazine
compliance-icons
Product Identifier
  • 3,4,5-Trichloropyridazine, 98%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'