Läs mer
3-[(4-Amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-5-[2-(sulfonatooxy)ethyl]thiazolium Chloride (Thiamine Chloride Sulfate Ester), Mikromol™
Beskrivning
3-[(4-Amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-5-[2-(sulfonatooxy)ethyl]thiazolium Chloride (Thiamine Chloride Sulfate Ester), Mikromol
Specifikationer
Specifikationer
| Kemiskt namn eller material | 3-[(4-Amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-5-[2-(sulfonatooxy)ethyl]thiazolium Chloride (Thiamine Chloride Sulfate Ester) |
| CAS | 15743-04-1 |
| Analyt- eller komponentnamn | 3-[(4-Amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-5-[2-(sulfonatooxy)ethyl]thiazolium Chloride (Thiamine Chloride Sulfate Ester) |
| Frakt skick | Room Temperature |
| Molekylformel | C12 H17 N4 O4 S2 . Cl |
| Certifikat och efterlevnad | ISO 17025 |
| Synonym | Thiamine Hydrochloride Imp. A (EP), Thiamine Imp. A (EP), 3-[(4-Amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-5-[2-(sulfonatooxy)ethyl]thiazolium Chloride, Thiamine Hydrochloride Imp. A (EP) as Chloride, Thiamine Chloride Sulfate Ester, Thiamine Hydrochloride Impurity A as Chloride, Thiamine Impurity A as Chloride |
| InChI formel | InChI=1S/C12H16N4O4S2.ClH/c1-8-11(3-4-20-22(17,18)19)21-7-16(8)6-10-5-14-9(2)15-12(10)13;/h5,7H,3-4,6H2,1-2H3,(H2-,13,14,15,17,18,19);1H |
| Lösningstyp | Neat |
| LEDER | [Cl-].Cc1ncc(C[n+]2csc(CCOS(=O)(=O)O)c2C)c(N)n1 |
| Visa mer |