Läs mer
3-((4-Amino-2-methylpyrimidin-5-yl)methyl)-5-(2-((hydroxy(phosphonooxy)phosphoryl)oxy)ethyl)-2-(1-hydroxyethyl)-4-methylthiazol-3-ium Chloride, TRC
Produktkod. 30326954
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30326954 10 mg
1 options
Produktkod. 30326954 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA20319010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-((4-Amino-2-methylpyrimidin-5-yl)methyl)-5-(2-((hydroxy(phosphonooxy)phosphoryl)oxy)ethyl)-2-(1-hydroxyethyl)-4-methylthiazol-3-ium Chloride
CAS 20319-27-1
Molekylformel C14 H23 N4 O8 P2 S . Cl
Kvantitet 10 mg
Synonym 2-(1-Hydroxyethyl)thiamine Pyrophosphate, 2-(α-Hydroxyethyl)thiamine Pyrophosphate
InChI formel InChI=1S/C14H22N4O8P2S.ClH/c1-8-12(4-5-25-28(23,24)26-27(20,21)22)29-14(9(2)19)18(8)7-11-6-16-10(3)17-13(11)15;/h6,9,19H,4-5,7H2,1-3H3,(H4-,15,16,17,20,21,22,23,24);1H
LEDER [Cl-].CC(O)c1sc(CCOP(=O)(O)OP(=O)(O)O)c(C)[n+]1Cc2cnc(C)nc2N
IUPAC-namn 2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-2-(1-hydroxyethyl)-4-methyl-1,3-thiazol-3-ium-5-yl]ethyl phosphono hydrogen phosphate;chloride
Molekylvikt (g/mol) 504.82
Formel vikt 504.04