Läs mer
3(4-Aminophenyl)-4-methyl-4,5-dihydro-1H-pyridazin-6-one, TRC
Produktkod. 30135034
Change view
Kvantitet:
1 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135034 25 g
30134383 10 g
17994925 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135034 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA62892025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 36725-28-7
Molekylformel C11 H13 N3 O
Molekylvikt (g/mol) 203.24
Synonym 6-(4-Aminophenyl)-4,5-dihydro-5-methyl-3(2H)-pyridazinone, 6-(p-Aminophenyl)-5-methyl-4,5-dihydro-3(2H)-pyridazinone, SKF 93505;
IUPAC-namn 3-(4-aminophenyl)-4-methyl-4,5-dihydro-1H-pyridazin-6-one
LEDER CC1CC(=O)NN=C1c2ccc(N)cc2
Specifikationer
Kemiskt namn eller material 3(4-Aminophenyl)-4-methyl-4,5-dihydro-1H-pyridazin-6-one
Analyt- eller komponentnamn 3(4-Aminophenyl)-4-methyl-4,5-dihydro-1H-pyridazin-6-one
Frakt skick Room Temperature
InChI formel InChI=1S/C11H13N3O/c1-7-6-10(15)13-14-11(7)8-2-4-9(12)5-3-8/h2-5,7H,6,12H2,1H3,(H,13,15)
Lösningstyp Neat
Industry Type Pharmaceutical
Kvantitet 25 g
Formel vikt 203.1059