Läs mer
3-(4-Biphenylyl)-1,2,3,4-tetrahydro-1-naphthol, TRC
Produktkod. 30341950
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30341950 500 mg
30348361 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30341950 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB398500500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-(4-Biphenylyl)-1,2,3,4-tetrahydro-1-naphthol
CAS 56181-66-9
Molekylformel C22H20O
Kvantitet 500 mg
Synonym 3-p-Biphenylyl-1-tetralinol, 3-[1,1'-Biphenyl]-4-yl-1,2,3,4-tetrahydro-1-naphthalenol
InChI formel InChI=1S/C22H20O/c23-22-15-20(14-19-8-4-5-9-21(19)22)18-12-10-17(11-13-18)16-6-2-1-3-7-16/h1-13,20,22-23H,14-15H2
LEDER OC1CC(Cc2ccccc12)c3ccc(cc3)c4ccccc4
IUPAC-namn 3-(4-phenylphenyl)-1,2,3,4-tetrahydronaphthalen-1-ol
Molekylvikt (g/mol) 300.39
Formel vikt 300.1514