missing translation for 'onlineSavingsMsg'
Läs mer

3,4-Bis(2-methoxyethoxy)benzoic Acid Ethyl Ester, Mikromol™

Produktkod. 30050116
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30050116 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30050116 Leverantör Mikromol Leverantörsnummer MM3297.090025

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

3,4-Bis(2-methoxyethoxy)benzoic Acid Ethyl Ester
TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material 3,4-Bis(2-methoxyethoxy)benzoic Acid Ethyl Ester
CAS 183322-16-9
Analyt- eller komponentnamn 3,4-Bis(2-methoxyethoxy)benzoic Acid Ethyl Ester
Frakt skick Room Temperature
Molekylformel C15 H22 O6
Certifikat och efterlevnad ISO 9001
Synonym Benzoic acid, 3,4-bis(2-methoxyethoxy)-, ethyl ester, Ethyl 3,4-bis(2-methoxyethoxy)benzoate
InChI formel InChI=1S/C15H22O6/c1-4-19-15(16)12-5-6-13(20-9-7-17-2)14(11-12)21-10-8-18-3/h5-6,11H,4,7-10H2,1-3H3
LEDER CCOC(=O)c1ccc(OCCOC)c(OCCOC)c1
IUPAC-namn ethyl 3,4-bis(2-methoxyethoxy)benzoate
Molekylvikt (g/mol) 298.33
Kvantitet 25 mg
Formel vikt 298.1416 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.