Läs mer
[3,4-Bis(benzyloxy)phenyl]acetic Acid Methyl Ester, TRC
Produktkod. 30329418
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30329418 250 mg
1 options
Produktkod. 30329418 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB416185250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material [3,4-Bis(benzyloxy)phenyl]acetic Acid Methyl Ester
CAS 15373-33-8
Molekylformel C23H22O4
Kvantitet 250 mg
Synonym 3,4-Bis(phenylmethoxy)benzeneacetic Acid Methyl Ester
InChI formel InChI=1S/C23H22O4/c1-25-23(24)15-20-12-13-21(26-16-18-8-4-2-5-9-18)22(14-20)27-17-19-10-6-3-7-11-19/h2-14H,15-17H2,1H3
LEDER O=C(CC1=CC=C(OCC2=CC=CC=C2)C(OCC3=CC=CC=C3)=C1)OC
IUPAC-namn methyl 2-(3,4-bis(benzyloxy)phenyl)acetate
Molekylvikt (g/mol) 362.42
Formel vikt 362.15