missing translation for 'onlineSavingsMsg'
Läs mer

3,4-Bis(etoxymetoxi)bensaldehyd, TRC

Product Code. 30333090
Change view
Click to view available options
Kvantitet:
2 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30333090 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30333090 Supplier Toronto Research Chemicals Supplier No. TRCB4336702G
 Begränsad artikel
Beräknad leverans: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Högrena organiska molekyler och analytiska standarder, strategiskt levererade världen över för att möjliggöra innovation och kommersiell framgång.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 3,4-Bis(ethoxymethoxy)benzaldehyde
CAS 128837-28-5
Molekylformel C13 H18 O5
Kvantitet 2 g
InChI formel InChI=1S/C13H18O5/c1-3-15-9-17-12-6-5-11(8-14)7-13(12)18-10-16-4-2/h5-8H,3-4,9-10H2,1-2H3
LEDER CCOCOc1ccc(C=O)cc1OCOCC
IUPAC-namn 3,4-bis(etoxymetoxi)bensaldehyd
Molekylvikt (g/mol) 254.28
Formel vikt 254.1154
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.