missing translation for 'onlineSavingsMsg'
Läs mer

3,4'-Bis(trifluoromethyl)benzhydrol, 97%, Thermo Scientific™

Produktkod. 15464227
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 g
5 g
Förpackningsstorlek:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
15464227 5 g 5g
15454227 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 15464227 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H31629.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 203915-48-4
Molekylformel C15H10F6O
Molekylvikt (g/mol) 320.23
MDL-nummer MFCD00045091
InChI-nyckel DEIGXBAZAOVYHC-UHFFFAOYNA-N
Synonym 3,4'-bis trifluoromethyl benzhydrol, 3-trifluoromethyl phenyl 4-trifluoromethyl phenyl methanol, benzenemethanol,3-trifluoromethyl-a-4-trifluoromethyl phenyl, 3-trifluoromethyl phenyl-4-trifluoromethyl phenyl methanol, bio1h11, 3,4-bis trifluoromethyl benzhydrol, 3,4'-bis trifluoromethyl phenylmethanol, bis 4-trifluoromethyl phenyl methan-1-ol
PubChem CID 2736114
IUPAC-namn [3-(trifluoromethyl)phenyl]-[4-(trifluoromethyl)phenyl]methanol
LEDER OC(C1=CC=C(C=C1)C(F)(F)F)C1=CC(=CC=C1)C(F)(F)F

Specifikationer

Kemiskt namn eller material 3,4'-Bis(trifluoromethyl)benzhydrol
Kvantitet 5 g
Formel vikt 320.23
Procent renhet 97%
Produkttitel
Välj ett problem

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.