Learn More
Alfa Aesar™ 3-(4-Bromophenyl)-2-thioxo-4-thiazolidinone, 95%
Brand:  Alfa Aesar™ H63281.03
Additional Details : CAS Number : 10574-70-6 Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15469898
  • 3114.20 SEK / 1g
Estimated Shipment date
from Supplier 06-08-2018
Add to basket
Specifications
Chemical Identifiers
10574-70-6
288.177
SQELHPVHPKNYGI-UHFFFAOYSA-N
99598
C9H6BrNOS2
MFCD00174856
3-(4-Bromophenyl)rhodanine
Specifications
3-(4-Bromophenyl)-2-thioxo-4-thiazolidinone
10574-70-6
Solid
C9H6BrNOS2
3-(4-Bromophenyl)rhodanine
C1C(=O)N(C(=S)S1)C2=CC=C(C=C2)Br
288.177
288.18
1g
95%
MFCD00174856
SQELHPVHPKNYGI-UHFFFAOYSA-N
3-(4-bromophenyl)-2-sulfanylidene-1,3-thiazolidin-4-one
99598