Läs mer
3-(4'-Carboxybenzylidene)camphor, TRC
Produktkod. 30349910
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30349910 500 mg
30351088 100 mg
30364888 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30349910 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC177806500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 68801-01-4
Molekylformel C18H20O3
Molekylvikt (g/mol) 284.35
Synonym 4-[(4,7,7-Trimethyl-3-oxobicyclo[2.2.1]hept-2-ylidene)methyl]benzoic Acid
IUPAC-namn 4-[(Z)-(4,7,7-trimethyl-3-oxo-2-bicyclo[2.2.1]heptanylidene)methyl]benzoic acid
LEDER CC1(C)C2CCC1(C)C(=O)/C/2=C\c3ccc(cc3)C(=O)O
Specifikationer
Kemiskt namn eller material 3-(4'-Carboxybenzylidene)camphor
Kvantitet 500 mg
InChI formel InChI=1S/C18H20O3/c1-17(2)14-8-9-18(17,3)15(19)13(14)10-11-4-6-12(7-5-11)16(20)21/h4-7,10,14H,8-9H2,1-3H3,(H,20,21)/b13-10-
Formel vikt 284.1412