Läs mer
3-(4-Chloro-2,6-dimethylphenyl)-8-methoxy-1-methyl-2-oxo-1,8-diazaspiro[4.5]dec-3-en-4-yl Ethyl Carbonate, TRC
Produktkod. 30367193
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30367193 10 mg
1 options
Produktkod. 30367193 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC23301010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Spiropidion
CAS 1229023-00-0
Molekylformel C21 H27 Cl N2 O5
Kvantitet 10 mg
Synonym Carbonic acid, 3-(4-chloro-2,6-dimethylphenyl)-8-methoxy-1-methyl-2-oxo-1,8-diazaspiro[4.5]dec-3-en-4-yl ethyl ester, 3-(4-chloro-2,6-dimethylphenyl)-8-methoxy-1-methyl-2-oxo-1,8-diazaspiro[4.5]dec-3-en-4-yl ethyl carbonate, Spiropidion, 3-(4-Chloro-2,6-dimethylphenyl)-8-methoxy-1-methyl-2-oxo-1,8-diazaspiro[4.5]dec-3-en-4-yl Ethyl Carbonate, Carbonic acid, 3-(4-chloro-2,6-dimethylphenyl)-8-methoxy-1-methyl-2-oxo-1,8-diazaspiro[4.5]dec-3-en-4-yl ethyl ester, 3-(4-chloro-2,6-dimethylphenyl)-8-methoxy-1-methyl-2-oxo-1,8-diazaspiro[4.5]dec-3-en-4-yl ethyl carbonate, Spiropidion
InChI formel InChI=1S/C21H27ClN2O5/c1-6-28-20(26)29-18-17(16-13(2)11-15(22)12-14(16)3)19(25)23(4)21(18)7-9-24(27-5)10-8-21/h11-12H,6-10H2,1-5H3
LEDER CCOC(=O)OC1=C(C(=O)N(C)C12CCN(CC2)OC)c3c(C)cc(Cl)cc3C
IUPAC-namn [3-(4-chloro-2,6-dimethylphenyl)-8-methoxy-1-methyl-2-oxo-1,8-diazaspiro[4.5]dec-3-en-4-yl] ethyl carbonate
Molekylvikt (g/mol) 422.902
Formel vikt 422.161