missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Chlorophenoxy)-1,2-propanediol, 99%, Thermo Scientific Chemicals

Catalog Number p-7023632
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
104-29-0
C9H11ClO3
202.634
MFCD00021990
MXOAEAUPQDYUQM-UHFFFAOYSA-N
chlorphenesin, 3-4-chlorophenoxy-1,2-propanediol, 3-4-chlorophenoxy propane-1,2-diol, chlorophenesin, chlorphenesinum, adermykon, gecophen, demykon, mycil, chlorphenesine
7697
CHEBI:3642
3-(4-chlorophenoxy)propane-1,2-diol
C1=CC(=CC=C1OCC(CO)O)Cl
This item is not returnable. View return policy
104-29-0
C9H11ClO3
202.634
MFCD00021990
MXOAEAUPQDYUQM-UHFFFAOYSA-N
chlorphenesin, 3-4-chlorophenoxy-1,2-propanediol, 3-4-chlorophenoxy propane-1,2-diol, chlorophenesin, chlorphenesinum, adermykon, gecophen, demykon, mycil, chlorphenesine
7697
CHEBI:3642
3-(4-chlorophenoxy)propane-1,2-diol
C1=CC(=CC=C1OCC(CO)O)Cl

It inhibits IgE-mediated histamine release. 3-(4-Chlorophenoxy)-1,2-propanediol is also used as an antimycotic agent. It is a centrally acting muscle relaxant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It inhibits IgE-mediated histamine release. 3-(4-Chlorophenoxy)-1,2-propanediol is also used as an antimycotic agent. It is a centrally acting muscle relaxant.

Solubility
Slightly soluble in water.

Notes
Store at 78-82°C. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 78°C to 82°C
Quantity 25 g
Solubility Information Slightly soluble in water.
Formula Weight 202.64
Percent Purity 99%
Chemical Name or Material 3-(4-Chlorophenoxy)-1,2-propanediol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'