missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(4-Chlorophenyl)-1-methyl-2-thiohydantoin, 95%

Brand:  Alfa Aesar™ H63731.06

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15490569

  • 7780.68 SEK / 5g


Estimated Shipment date
from Supplier 06-08-2018
Add to basket

Specifications

Chemical Identifiers

3-(4-Chlorophenyl)-1-methyl-2-thiohydantoin
MFCD03719914
3-(4-Chlorophenyl)-1-methyl-2-thioxo-4-imidazolidinone

Specifications

3-(4-Chlorophenyl)-1-methyl-2-thiohydantoin
5g
95%
3-(4-Chlorophenyl)-1-methyl-2-thioxo-4-imidazolidinone
240.71
Solid
MFCD03719914
Videos
SDS