Läs mer
3-(4-Chlorophenyl)glutaric Acid, Mikromol™
Produktkod. 17972966
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17972966 25 mg
17921126 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17972966 Leverantör Mikromol Leverantörsnummer MM1235.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3-(4-Chlorophenyl)glutaric Acid
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-(4-Chlorophenyl)glutaric Acid
CAS 35271-74-0
Analyt- eller komponentnamn 3-(4-Chlorophenyl)glutaric Acid
Frakt skick Room Temperature
Molekylformel C11 H11 Cl O4
Certifikat och efterlevnad ISO 17025
Synonym 3-(4-Chlorophenyl)glutaric Acid
InChI formel InChI=1S/C11H11ClO4/c12-9-3-1-7(2-4-9)8(5-10(13)14)6-11(15)16/h1-4,8H,5-6H2,(H,13,14)(H,15,16)
LEDER OC(=O)CC(CC(=O)O)c1ccc(Cl)cc1
IUPAC-namn 3-(4-chlorophenyl)pentanedioic acid
Molekylvikt (g/mol) 242.66
Kvantitet 25 mg
Formel vikt 242.0346 g/mol