missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-[(4-Chlorophenylsulfonyl)methyl]benzamidoxime, Alfa Aesar™
Click to view available options
Quantity:
1 g
5 g
Specifications
Quantity | 1 g |
Formula Weight | 324.79 |
Chemical Name or Material | 3-[(4-Chlorophenylsulfonyl)methyl]benzamidoxime |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'