Läs mer
3,4-Di-O-benzyl DL-threo-Droxidopa Hydrochloride, TRC
Produktkod. 30352734
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30352734 25 mg
1 options
Produktkod. 30352734 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD41753025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3,4-Di-O-benzyl DL-threo-Droxidopa Hydrochloride
CAS 73594-43-1
Molekylformel C23H24ClNO5
Kvantitet 25 mg
Synonym DL-threo-3-[3,4-Bis(benzyloxy)phenyl)serine Hydrochloride, beta-Hydroxy-3-(phenylmethoxy)-O-(phenylmethyl)-threo-DL-tyrosine Hydrochloride
InChI formel InChI=1S/C23H23NO5.ClH/c24-21(23(26)27)22(25)18-11-12-19(28-14-16-7-3-1-4-8-16)20(13-18)29-15-17-9-5-2-6-10-17;/h1-13,21-22,25H,14-15,24H2,(H,26,27);1H/t21-,22+;/m0./s1
LEDER Cl.N[C@@H]([C@H](O)c1ccc(OCc2ccccc2)c(OCc3ccccc3)c1)C(=O)O
IUPAC-namn (2S,3R)-2-amino-3-[3,4-bis(phenylmethoxy)phenyl]-3-hydroxypropanoic acid;hydrochloride
Molekylvikt (g/mol) 429.89
Formel vikt 429.1343