Läs mer
3,4-Di-O-benzyl Droxidopa Hydrochloride(Mixture of Diastereomers), TRC
Produktkod. 30361135
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30361135 50 mg
1 options
Produktkod. 30361135 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD41752550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3,4-Di-O-benzyl Droxidopa Hydrochloride(Mixture of Diastereomers)
CAS 1323257-61-9
Molekylformel C23H24ClNO5
Kvantitet 50 mg
Synonym beta-Hydroxy-3-(phenylmethoxy)-O-(benzyl)tyrosine, beta-Hydroxy-3-(phenylmethoxy)-O-(phenylmethyl)tyrosine
InChI formel InChI=1S/C23H23NO5.ClH/c24-21(23(26)27)22(25)18-11-12-19(28-14-16-7-3-1-4-8-16)20(13-18)29-15-17-9-5-2-6-10-17;/h1-13,21-22,25H,14-15,24H2,(H,26,27);1H
LEDER Cl.NC(C(O)c1ccc(OCc2ccccc2)c(OCc3ccccc3)c1)C(=O)O
IUPAC-namn 2-amino-3-[3,4-bis(phenylmethoxy)phenyl]-3-hydroxypropanoic acid;hydrochloride
Molekylvikt (g/mol) 429.89
Formel vikt 429.1343