missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3,4-Diamino-2-methylpyrazole sulfate, 95%

Brand:  Alfa Aesar™ H61986.03

Additional Details : CAS Number : 20055-01-0 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15468578

  • 778.07 SEK / 1g


Estimated Shipment date
from Supplier 04-07-2018
Add to basket

Specifications

Chemical Identifiers

3,4-Diamino-2-methylpyrazole sulfate
20055-01-0
210.21

Specifications

3,4-Diamino-2-methylpyrazole sulfate
20055-01-0
95%
1g
Solid
210.21
Videos
SDS