Learn More
3,4-Diaminoanisole dihydrochloride, 98%, Thermo Scientific Chemicals
Catalog Number p-4812878
Click to view available options
Quantity:
1 g
5 g
25 g
59548-39-9
C7H10N22ClH
211.09
SXCHMHOBHJOXGC-UHFFFAOYSA-N
4-methoxybenzene-1,2-diamine dihydrochloride, 4-methoxy-o-phenylenediamine dihydrochloride, 3,4-diaminoanisole dihydrochloride, 1,2-benzenediamine, 4-methoxy-, dihydrochloride, 3,4-diaminoanisole 2hcl, 4-methoxy-phenylenediamine dihydrochloride, 4-methoxy-1,2-benzenediamine dihydrochloride, 4-methoxy-benzene-1,2-diamine dihydrochloride, 4-methoxy-1,2-phenylenediamine dihydrochloride, 4-methoxy-l,2-phenylene diamine dihydrochloride
3085161
4-methoxybenzene-1,2-diamine;dihydrochloride
COC1=CC(=C(C=C1)N)N.Cl.Cl
59548-39-9
C7H10N22ClH
211.09
SXCHMHOBHJOXGC-UHFFFAOYSA-N
4-methoxybenzene-1,2-diamine dihydrochloride, 4-methoxy-o-phenylenediamine dihydrochloride, 3,4-diaminoanisole dihydrochloride, 1,2-benzenediamine, 4-methoxy-, dihydrochloride, 3,4-diaminoanisole 2hcl, 4-methoxy-phenylenediamine dihydrochloride, 4-methoxy-1,2-benzenediamine dihydrochloride, 4-methoxy-benzene-1,2-diamine dihydrochloride, 4-methoxy-1,2-phenylenediamine dihydrochloride, 4-methoxy-l,2-phenylene diamine dihydrochloride
3085161
4-methoxybenzene-1,2-diamine;dihydrochloride
COC1=CC(=C(C=C1)N)N.Cl.Cl
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 206.0°C to 209.0°C
Assay Percent Range 97.5 to max. 102.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula CH3OC6H3(NH2)2·2HCl
Quantity 1 g
Solubility Information Solubility in water: soluble.
Formula Weight 211.09
Percent Purity 98%
Chemical Name or Material 3, 4-Diaminoanisole dihydrochloride
compliance-icons
Product Identifier
  • 3,4-Diaminoanisole dihydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.