Läs mer
3,4-Diaminobenzonitrile, TRC
Produktkod. 30350745
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30350745 1 g
30361330 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30350745 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4162201G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3,4-Diaminobenzonitrile
CAS 17626-40-3
Molekylformel C7 H7 N3
Kvantitet 1 g
InChI formel InChI=1S/C7H7N3/c8-4-5-1-2-6(9)7(10)3-5/h1-3H,9-10H2
LEDER Nc1ccc(cc1N)C#N
IUPAC-namn 3,4-diaminobenzonitrile
Molekylvikt (g/mol) 133.15
Formel vikt 133.064