Läs mer
3,4-Diaminobenzophenone, TRC
Produktkod. 30358610
Change view
Kvantitet:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30358610 10 g
30350594 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30358610 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD41610010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3,4-Diaminobenzophenone
CAS 39070-63-8
Molekylformel C13H12N2O
Kvantitet 10 g
Synonym (3,4-Diaminophenyl)phenylmethanone, 1,2-Diamino-4-(phenylcarbonyl)benzene, 2-Amino-4-benzoylaniline, 3,4- Benzophenonediamine, 3,4-Diaminobenzophenone, 4-Benzoyl-1,2-benzenediamine, 4-Benzoyl-1,2-diaminobenzene, 4-Benzoyl-1,2- phenylenediamine, 4-Benzoyl-o-phenylenediamine, p-Benzoyl-o-phenylenediamine
InChI formel InChI=1S/C13H12N2O/c14-11-7-6-10(8-12(11)15)13(16)9-4-2-1-3-5-9/h1-8H,14-15H2
LEDER O=C(C1=CC=CC=C1)C2=CC=C(N)C(N)=C2
IUPAC-namn (3,4-diaminophenyl)(phenyl)methanone
Molekylvikt (g/mol) 212.25
Formel vikt 212.09