Läs mer
3,4-Diaminophenol (>80%), TRC
Produktkod. 30358962
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30358962 50 mg
30355721 500 mg
30353556 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30358962 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD41664850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 615-72-5
Molekylformel C6H8N2O
Molekylvikt (g/mol) 124.14
Synonym 3,4-Diaminophenol
IUPAC-namn 3,4-diaminophenol
LEDER Nc1ccc(O)cc1N
Specifikationer
Kemiskt namn eller material 3,4-Diaminophenol
Kvantitet 50 mg
InChI formel InChI=1S/C6H8N2O/c7-5-2-1-4(9)3-6(5)8/h1-3,9H,7-8H2
Formel vikt 124.0637