Läs mer
3,4-Dibenzyl-gallic Acid Benzyl Ester, TRC
Produktkod. 30366031
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30366031 50 mg
1 options
Produktkod. 30366031 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD41798550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3,4-Dibenzyl-gallic Acid Benzyl Ester
CAS 1159977-28-2
Molekylformel C28H24O5
Kvantitet 50 mg
Synonym Benzyl 3,4-Di-O-benzyl-5-hydroxy Benzoate, 3,4-Diphenylmethoxy-5-hydroxy-benzoic Acid Benzyl Ester, Benzyl 3,4-Dibenzyloxy-5-hydroxy Benzoate, 3-Hydroxy-4,5-bis(phenylmethoxy)benzoic Acid Phenylmethyl Ester
InChI formel InChI=1S/C28H24O5/c29-25-16-24(28(30)33-20-23-14-8-3-9-15-23)17-26(31-18-21-10-4-1-5-11-21)27(25)32-19-22-12-6-2-7-13-22/h1-17,29H,18-20H2
LEDER Oc1cc(cc(OCc2ccccc2)c1OCc3ccccc3)C(=O)OCc4ccccc4
IUPAC-namn benzyl 3-hydroxy-4,5-bis(phenylmethoxy)benzoate
Molekylvikt (g/mol) 440.49
Formel vikt 440.1624