missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichloro-1,2,5-thiadiazole, 99%, Thermo Scientific Chemicals

Catalog Number p-7081668
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
5728-20-1
C2Cl2N2S
155.00
MFCD00010072
YNZQOVYRCBAMEU-UHFFFAOYSA-N
1,2,5-thiadiazole, 3,4-dichloro, dichloro-1,2,5-thiadiazole, 3,4-dichloro-1,2,5 thiadiazole, pubchem20790, acmc-1al9l, ksc272e3d, 4,5-dichloro-2,1,3-thiadiazole, 3,4-bis chloranyl-1,2,5-thiadiazole, 3,4-dichloro-1,2,5-thiadiazole
79804
3,4-dichloro-1,2,5-thiadiazole
ClC1=NSN=C1Cl
This item is not returnable. View return policy
5728-20-1
C2Cl2N2S
155.00
MFCD00010072
YNZQOVYRCBAMEU-UHFFFAOYSA-N
1,2,5-thiadiazole, 3,4-dichloro, dichloro-1,2,5-thiadiazole, 3,4-dichloro-1,2,5 thiadiazole, pubchem20790, acmc-1al9l, ksc272e3d, 4,5-dichloro-2,1,3-thiadiazole, 3,4-bis chloranyl-1,2,5-thiadiazole, 3,4-dichloro-1,2,5-thiadiazole
79804
3,4-dichloro-1,2,5-thiadiazole
ClC1=NSN=C1Cl

3,4-Dichloro-1,2,5-thiadiazole is a chlorinated thiadiazole that is a potent inhibitors of nitrification in soil. 3,4-Dichloro-1,2,5-thiadiazole is used as a building block in the preparaton of the β-adrenergic blocking agent, Timolol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dichloro-1,2,5-thiadiazole is a chlorinated thiadiazole that is a potent inhibitors of nitrification in soil. 3,4-Dichloro-1,2,5-thiadiazole is used as a building block in the preparaton of the β-adrenergic blocking agent, Timolol.

Solubility
Soluble in chloroform and methanol.

Notes
Hygroscopic. Store under dry inert gas. Incompatible with moisture and oxidizing agents, acids and bases.
TRUSTED_SUSTAINABILITY
Density 1.644
Boiling Point 157°C to 159°C
Flash Point >100°C (212°F)
Refractive Index 1.562
Quantity 10 g
UN Number UN2810
Beilstein 606585
Sensitivity Hygroscopic
Solubility Information Soluble in chloroform and methanol.
Formula Weight 155.01
Percent Purity 99%
Chemical Name or Material 3,4-Dichloro-1,2,5-thiadiazole
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'