missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichloro-L-phenylalanine, 98%, Thermo Scientific Chemicals

Catalog Number p-7055298
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
52794-99-7
C9H9Cl2NO2
234.076
MFCD00191203
NRCSJHVDTAAISV-QMMMGPOBSA-N
s-2-amino-3-3,4-dichlorophenyl propanoic acid, l-3,4-dichlorophenylalanine, 3,4-dichloro-l-phenylalanine, 2s-2-amino-3-3,4-dichlorophenyl propanoic acid, h-phe 3,4-dicl-oh, 3,4-dichloro-phenylalanine, l-3,4-dichlorophenlalanine, l-3,4-dichlorophe, h-3,4-dichloro-phe-oh, dichlorophe
2761498
(2S)-2-amino-3-(3,4-dichlorophenyl)propanoic acid
C1=CC(=C(C=C1CC(C(=O)O)N)Cl)Cl
This item is not returnable. View return policy
52794-99-7
C9H9Cl2NO2
234.076
MFCD00191203
NRCSJHVDTAAISV-QMMMGPOBSA-N
s-2-amino-3-3,4-dichlorophenyl propanoic acid, l-3,4-dichlorophenylalanine, 3,4-dichloro-l-phenylalanine, 2s-2-amino-3-3,4-dichlorophenyl propanoic acid, h-phe 3,4-dicl-oh, 3,4-dichloro-phenylalanine, l-3,4-dichlorophenlalanine, l-3,4-dichlorophe, h-3,4-dichloro-phe-oh, dichlorophe
2761498
(2S)-2-amino-3-(3,4-dichlorophenyl)propanoic acid
C1=CC(=C(C=C1CC(C(=O)O)N)Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Formula Weight 234.08
Percent Purity 98%
Quantity 1 g
Physical Form Solid
Chemical Name or Material 3,4-Dichloro-L-phenylalanine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'