Läs mer
3,4-Didehydro Naratriptan (Naratriptan Impurity B), TRC
Produktkod. 30135082
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135082 100 mg
30132426 25 mg
30008112 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135082 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD449205100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 121679-20-7
Molekylformel C17 H23 N3 O2 S
Molekylvikt (g/mol) 333.45
Synonym 3,4-Dihydro Naratriptan (Naratriptan Impurity B), N-Methyl-3-(1,2,3,6-tetrahydro-1-methyl-4-pyridinyl)-1H-indole-5-ethanesulfonamide
IUPAC-namn N-methyl-2-[3-(1-methyl-3,6-dihydro-2H-pyridin-4-yl)-1H-indol-5-yl]ethanesulfonamide
LEDER CNS(=O)(=O)CCc1ccc2[nH]cc(C3=CCN(C)CC3)c2c1
Specifikationer
Kemiskt namn eller material 3,4-Dihydro Naratriptan (Naratriptan Impurity B)
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C17H23N3O2S/c1-18-23(21,22)10-7-13-3-4-17-15(11-13)16(12-19-17)14-5-8-20(2)9-6-14/h3-5,11-12,18-19H,6-10H2,1-2H3
Formel vikt 333.1511
Procent renhet >95%