Learn More
Alfa Aesar™ 3,4-Difluoro-N-(2-methylbenzyl)benzenesulfonamide, 97%
Brand:  Alfa Aesar™ H61484.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15486546
  • 5659.56 SEK / 1g
Estimated Shipment date
from Supplier 05-03-2018
Add to basket
Specifications
Chemical Identifiers
3,4-Difluoro-N-(2-methylbenzyl)benzenesulfonamide
297.32
MFCD15533413
Specifications
3,4-Difluoro-N-(2-methylbenzyl)benzenesulfonamide
Solid
297.32
1g
97%
MFCD15533413